Learn More
Medicus Health Pipette, Universal Safety Filler Bulb, 3 PK
SDP
Catalog No. NC3443511
Encompass
Pipette, Universal Safety Filler Bulb features a silicone conical adapter that creates a tight seal without twisting
∙ Pack of 3 Each
∙ Pipette doesn't come in contact with hard plastic, thus reducing the risk of cracking and chipping
∙ 15mL capacity, 106mm L x 41mm OD (4.5-6.5mm hole dia.)
∙ Use the supplied elastic band for hanging on reagent bottles or wrist
∙ This pipette filler is for use with standard size pipettes up to 25mL in capacity
∙ 4.2" L
Catalog No. NC3443511 Supplier Medicus Health Supplier No. 2914M1
Pipette, Universal Safety Filler Bulb features a silicone conical adapter that creates a tight seal without twisting
∙ Pack of 3 Each
∙ Pipette doesn't come in contact with hard plastic, thus reducing the risk of cracking and chipping
∙ 15mL capacity, 106mm L x 41mm OD (4.5-6.5mm hole dia.)
∙ Use the supplied elastic band for hanging on reagent bottles or wrist
∙ This pipette filler is for use with standard size pipettes up to 25mL in capacity
∙ 4.2" L