Learn More
Electron Microscopy Sciences Bench Top Pipette Workstation
SDP
Catalog No. 5030592
Encompass_Preferred
This workstation allows you to keep the pipettor and the disposable tips handy at all times in one convenient box. The built-in rack stores up to 6 pipettors and the self-closing lid provides dust-free storage. The bottom, sides, and top pipettor rack are made from white acrylic and the two bins have a transparent front for easy identification of their contents.
Catalog No. 50-305-92 Supplier Electron Microscopy Sciences Supplier No. 6437401
This workstation allows you to keep the pipettor and the disposable tips handy at all times in one convenient box. The built-in rack stores up to 6 pipettors and the self-closing lid provides dust-free storage. The bottom, sides, and top pipettor rack are made from white acrylic and the two bins have a transparent front for easy identification of their contents.