Learn More
Electron Microscopy Sciences UV Stable Adjustable Pipetters, 1000 mcL- 5000 mcL, 10.0 µl Increment
SDP
Catalog No. 501899899
Encompass_Preferred
UV stable adjustable pipetters are autoclavable with no need to recalibrate after autoclaving. Highly resistant to chemical reagents. Barrel is designed to minimized dead air space for increased pipetter accuracy. The lightweight, slender, design reduces operator fatigue.
Range: 1000 µl- 5000 µl
Catalog No. 50-189-9899 Supplier Electron Microscopy Sciences Supplier No. 7243014
UV stable adjustable pipetters are autoclavable with no need to recalibrate after autoclaving. Highly resistant to chemical reagents. Barrel is designed to minimized dead air space for increased pipetter accuracy. The lightweight, slender, design reduces operator fatigue.
Range: 1000 µl- 5000 µl