Learn More
Electron Microscopy Sciences UV Stable Adjustable Pipetters, 20 mcL- 200 mcL, 1.0 µl Increment
SDP
Catalog No. 501899897
Encompass_Preferred
UV stable adjustable pipetters are autoclavable with no need to recalibrate after autoclaving. Highly resistant to chemical reagents. Barrel is designed to minimized dead air space for increased pipetter accuracy. The lightweight, slender, design reduces operator fatigue.
Range: 20 µl- 200 µl
Catalog No. 50-189-9897 Supplier Electron Microscopy Sciences Supplier No. 7243010
UV stable adjustable pipetters are autoclavable with no need to recalibrate after autoclaving. Highly resistant to chemical reagents. Barrel is designed to minimized dead air space for increased pipetter accuracy. The lightweight, slender, design reduces operator fatigue.
Range: 20 µl- 200 µl