Learn More
Electron Microscopy Sciences UV Stable Adjustable Pipetters, 2 mcL-20 mcL, 0.1 µl Increment
SDP
Catalog No. 501899895
Encompass_Preferred
UV stable adjustable pipetters are autoclavable with no need to recalibrate after autoclaving. Highly resistant to chemical reagents. Barrel is designed to minimized dead air space for increased pipetter accuracy. The lightweight, slender, design reduces operator fatigue.
Range: 2 µl-20 µl
Catalog No. 50-189-9895 Supplier Electron Microscopy Sciences Supplier No. 7243006
UV stable adjustable pipetters are autoclavable with no need to recalibrate after autoclaving. Highly resistant to chemical reagents. Barrel is designed to minimized dead air space for increased pipetter accuracy. The lightweight, slender, design reduces operator fatigue.
Range: 2 µl-20 µl