Learn More
Electron Microscopy Sciences UV Stable Adjustable Pipetters, 2 mcL- 100 mcL, 0.1 µl Increment
SDP
Catalog No. 501899896
Encompass_Preferred
UV stable adjustable pipetters are autoclavable with no need to recalibrate after autoclaving. Highly resistant to chemical reagents. Barrel is designed to minimized dead air space for increased pipetter accuracy. The lightweight, slender, design reduces operator fatigue.
Range: 2 µl- 100 µl
Catalog No. 50-189-9896 Supplier Electron Microscopy Sciences Supplier No. 7243008
UV stable adjustable pipetters are autoclavable with no need to recalibrate after autoclaving. Highly resistant to chemical reagents. Barrel is designed to minimized dead air space for increased pipetter accuracy. The lightweight, slender, design reduces operator fatigue.
Range: 2 µl- 100 µl