Learn More
Andwin Scientific PIPETTER BASIN 55 ML
SDP
Catalog No. NC0249354
Encompass_Preferred
The 55 mL pipettor basin is a disposable item that is compatible to use with 8 and 12 channel pipettors. With a capacity of 55 mL and a "V" shaped bottom, this product provides almost complete solution withdrawal. It is made of polyvinyl chloride material that permits the contents to be clearly viewed. This product is sold in a pack of 100 basins measuring 14.3 x 7 x 2.5 cm (55/8 x 23/4 x 1").
Catalog No. NC0249354 Supplier Andwin Scientific Supplier No. 21007970
The 55 mL pipettor basin is a disposable item that is compatible to use with 8 and 12 channel pipettors. With a capacity of 55 mL and a "V" shaped bottom, this product provides almost complete solution withdrawal. It is made of polyvinyl chloride material that permits the contents to be clearly viewed. This product is sold in a pack of 100 basins measuring 14.3 x 7 x 2.5 cm (55/8 x 23/4 x 1").