Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies Pipetter Nozzles
GSA_VASDP

Catalog No. 22146660 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Volume (Metric):
0.5 to 20 μL
100 to 200 μL
2 μL
2 to 20 μL
5 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-146-660 100 to 200 μL
22-146-653 2 to 20 μL
22-146-656 2 μL
22-146-244 0.5 to 20 μL
22-146-306 5 mL
22-146-664 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-146-660 Supplier VistaLab™ Technologies Supplier No. 90572010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains an internal aerosol/liquid barrier filter to prevent liquid from being aspirated into pipetter

  • Available in range of sizes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

For Use With Pipettes
Material Plastic
Sterility Non-sterile
Volume (Metric) 100 to 200 μL
For Use With (Equipment) 2 to 125 μL Electronic Single Channel Model, 5 to 250 μL and 20 to 200 μL Adjustable Model, 21 to 200 μL Fixed Models
Includes Internal aerosol/liquid barrier filter
Quantity 10/Box
Length (English) 5 in.
Width (English) 3 in.
Height (English) 1 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.