Learn More
World Precision Instrument Universal Filter Pipetter Tips-500200
SDP
Catalog No. 50822086
Encompass
WPI’s universal Ultra-clear, certified RNase/DNase-free polypropylene pipette tips are for use with Regal and most other pipetters, including Gilson, Oxford Benchmate, Socorex, and SealPette.
Catalog No. 50-822-086 Supplier World Precision Instrument Supplier No. 500200
WPI’s universal Ultra-clear, certified RNase/DNase-free polypropylene pipette tips are for use with Regal and most other pipetters, including Gilson, Oxford Benchmate, Socorex, and SealPette.