Learn More
Andwin Scientific BD Falcon Disposable Serological Pipets Polystyrene Plugged Sterile 50mL. Increments: 1.0mL
SDP
Catalog No. NC9003164
Encompass_Preferred
BD Falcon Disposable Serological Pipets Polystyrene Plugged Sterile 50mL. Increments: 1.0mL. Overfill capacity: 10.0mL. Color code: Black. Reverse graduations. Graduation alignment mark. Certified nonpyrogenic. Individually wrapped in paper.
Catalog No. NC9003164 Supplier Andwin Scientific Supplier No. 53106441
BD Falcon Disposable Serological Pipets Polystyrene Plugged Sterile 50mL. Increments: 1.0mL. Overfill capacity: 10.0mL. Color code: Black. Reverse graduations. Graduation alignment mark. Certified nonpyrogenic. Individually wrapped in paper.