Learn More
Haztech Systems, Inc Hazcat Plastic Pipettes. 2ml Graduated. Box of 500
SDP
Catalog No. NC9753350
Encompass
Hazcat polyethylene transfer pipettes. These pipettes are the correct size to make gas collection bulbs when used with the Hazcat System. Graduated to 2ml. Box of 500ea.
Catalog No. NC9753350 Supplier Haztech Systems, Inc Supplier No. HW4050
Hazcat polyethylene transfer pipettes. These pipettes are the correct size to make gas collection bulbs when used with the Hazcat System. Graduated to 2ml. Box of 500ea.