Learn More
Electron Microscopy Sciences Pipettes Volumetric 1 ml, 0.1 ml gradations
SDP
Catalog No. 501899377
Encompass_Preferred
Made from translucent polypropylene. Resistant to breakage. Blue graduations, ring mark and inscriptions. Ideal for fluoro and silicate chemistry used. Recommended cleaning with mild alkaline detergent up to 60 degree C. NOTE: Exposure to temperatures above 60 degree C may cause volume changes.
Capacity: 1 ml
Gradations: 0.1 ml
Catalog No. 50-189-9377 Supplier Electron Microscopy Sciences Supplier No. 7095201
Made from translucent polypropylene. Resistant to breakage. Blue graduations, ring mark and inscriptions. Ideal for fluoro and silicate chemistry used. Recommended cleaning with mild alkaline detergent up to 60 degree C. NOTE: Exposure to temperatures above 60 degree C may cause volume changes.
Capacity: 1 ml
Gradations: 0.1 ml