Learn More
Electron Microscopy Sciences Exact Volume Pipet 40 µl, 2.3 mm, 5.9 cm Stem diameter
SDP
Catalog No. 50365803
Encompass_Preferred
These pipets offer the exact volume of liquid just one squeeze to the bulb. Quick and economical for disposable pipets, without the need of expensive pipetting devices. These pipets come non-sterile.
Volume: 2.3 mm
Stem diameter: 5.9 cm
OAL: Bulk
Catalog No. 50-365-803 Supplier Electron Microscopy Sciences Supplier No. 7096933
These pipets offer the exact volume of liquid just one squeeze to the bulb. Quick and economical for disposable pipets, without the need of expensive pipetting devices. These pipets come non-sterile.
Volume: 2.3 mm
Stem diameter: 5.9 cm
OAL: Bulk