Learn More
Chemglass Life Sciences Pipette Tip, 10µL, Standard, Barrier Tip, Sterile, 960/Case
SDP
Catalog No. 501532326
Encompass_Preferred
Pipette Tip, 10¿L, Standard, Barrier Tip, Sterile, 960/Case. Filtered pipette tips are the experienced scientists choice for eliminating carryover contamination in the laboratory environment. Chemglass's market leading, smooth surface technology helps facilitate the release of the tip from the mold, eliminating the need for releasing fluids which can cause contamination. Chemglass's aerosol barrier filters are molded with ultra-high molecular weight polyethylene, completely inert and consistent in porosity. This ensures that the filter/barrier within the tips will not impede airflow and will protect your samples and pipettor from harmful contamination. The filters are manufactured within a clean-room facility, therefore every component is free of nucleic acid and enzymes which are detrimental to your research. All the filtered tips are pre-sterilized and certified by an independent lab to be free of nucleic acids, RNase, DNase and pyrogens.
Catalog No. 50-153-2326 Supplier Chemglass Life Sciences Supplier No. CLS3499C018
Pipette Tip, 10¿L, Standard, Barrier Tip, Sterile, 960/Case. Filtered pipette tips are the experienced scientists choice for eliminating carryover contamination in the laboratory environment. Chemglass's market leading, smooth surface technology helps facilitate the release of the tip from the mold, eliminating the need for releasing fluids which can cause contamination. Chemglass's aerosol barrier filters are molded with ultra-high molecular weight polyethylene, completely inert and consistent in porosity. This ensures that the filter/barrier within the tips will not impede airflow and will protect your samples and pipettor from harmful contamination. The filters are manufactured within a clean-room facility, therefore every component is free of nucleic acid and enzymes which are detrimental to your research. All the filtered tips are pre-sterilized and certified by an independent lab to be free of nucleic acids, RNase, DNase and pyrogens.