Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Pipetting By Numbers: STEAM Pipetting Practice Kit
SDPSCI_ED

Catalog No. S27559 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S27559 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27559 Supplier Edvotek™ Supplier No. S45
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Helps students master the fundamental biotechnology technique of micropipetting while creating a colorful, science-themed classroom poster with their own artwork

  • Storage: Room Temperature
  • Completion Time: 45 Minutes
  • For 10 Lab Groups

Specifications

Grade 7 to Undergraduate
Age 12 to 20
Includes Instructions, Dyes, Test Strips, Pipetting By Numbers Canvas, Blank Canvases, and Microcentrifuge Tubes
Requires Automatic Micropipettes with Tips, Small Containers for Discarding Tips, Scissors, and Tape
Time Range 45 min.
Content And Storage Room Temperature
Class Size 10 Lab Groups
Product Type Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.