Learn More
Cole-Parmer Rubber Bulb Pipette Filler, 60 mL capacity
Catalog No. NC0236962 Shop All Cole-Parmer Products
Encompass_Preferred
Rubber Bulb Pipette Filler, 60 mL capacityThese natural rubber bulbs can be used with Pasteur pipettes and small pipettes. Bulbs have a thickened wall to grip the glass firmly and protect it.Overall Length (in)3.25
Catalog No. NC0236962 Supplier Cole-Parmer Supplier No. 2481060
Rubber Bulb Pipette Filler, 60 mL capacityThese natural rubber bulbs can be used with Pasteur pipettes and small pipettes. Bulbs have a thickened wall to grip the glass firmly and protect it.Overall Length (in)3.25