Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Pipettor Tips Pack
SDP

Catalog No. 13299568
Change view
Click to view available options
Quantity:
30 Pipette Tips/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-299-568 30 Pipette Tips/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-568 Supplier CHEMetrics™ by AquaPhoenix Supplier No. A0171
Add to Cart
Add to Cart

Pack of pipettor tips for laboratory and testing applications. Includes 30 pipette tips per pack.

  • Each A-0171 pipette tip is designed to be used only once.

Specifications

Filtered Non-filtered
Sterility Non-sterile
Color Yellow
Autoclavable Not Autoclavable
Format Bagged
Tip Style Standard
Quantity 30 Pipette Tips/Pk.
Product Type Pipettor Tip
For Use With (Equipment) Universal Fit
Material Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.