Learn More
Aviva Systems Biology Corp PIR Antibody - C-terminal region (ARP33664_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33664_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PIR. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human PIR. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55109