Learn More
Medchemexpress LLC HY-B0585 500mg Medchemexpress, Piracetam CAS:7491-74-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0585500MG
Encompass_Preferred
Medchemexpress, HY-B0585 500mg Piracetam CAS:7491-74-9 Piracetam (UCB-6215) is a cyclic derivative of the neurotransmitter gamma-aminobutyric acid (GABA), used in treatment of a wide range of cognitive disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2610