Learn More
C&G Containers PIRAMAL 2OZ CBR WPTFESPTC288CS
SDP
Catalog No. NC0434111
Encompass_Preferred
2oz Flint glass Piramal Boston Round 20-410 White 2-piece Hole Cap w/PTFE septa, assembled only (bottle with cap attached) L3 288/cs
Catalog No. NC0434111 Supplier C&G Containers Supplier No. LFB002901600
2oz Flint glass Piramal Boston Round 20-410 White 2-piece Hole Cap w/PTFE septa, assembled only (bottle with cap attached) L3 288/cs