Learn More
Apexbio Technology LLC Pirarubicin, 10mg. Cas: 72496-41-4 MFCD: MFCD00869742. Topoisomerase II inhibitor;anthracycline antibiotic
SDP
Supplier:  Apexbio Technology LLC B229510
Encompass_Preferred
Pirarubicin is an analogue of the anthracycline anti-neoplastic doxorubicin. Intercalates into DNA and interacts with Topo II (topoisomerase II) and supressing DNA replication. Pirarubicin is an inhibitor of Topo II. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5340