Learn More
Medchemexpress LLC HY-13725 10mg Medchemexpress, Pirarubicin CAS:72496-41-4 Purity:>98%
SDP
Catalog No. 502081481
Encompass_Preferred
Medchemexpress, HY-13725 10mg Pirarubicin CAS:72496-41-4 Pirarubicin is an anthracycline antibiotics, acts as a topoisomerase II inhibitor, and is a widely used for treatment of various cancers, in particular, solid tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1481 Supplier Medchemexpress LLC Supplier No. HY1372510MG
Medchemexpress, HY-13725 10mg Pirarubicin CAS:72496-41-4 Pirarubicin is an anthracycline antibiotics, acts as a topoisomerase II inhibitor, and is a widely used for treatment of various cancers, in particular, solid tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.