Learn More
Apexbio Technology LLC Pirarubicin 72496-41-4 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B22955.1
Encompass_Preferred
Pirarubicin is a DNA intercalating anthracycline antibiotic structurally related to doxorubicin that targets the DNA topoisomerase II enzyme By stabilizing the cleavable DNA-enzyme complex pirarubicin induces DNA strand breaks and inhibits DNA replication and cellular proliferation Its primary mode of action involves disruptive interactions with topoisomerase II-mediated DNA cleavage and repair mechanisms Pirarubicin is utilized as an antineoplastic agent in oncology research particularly in evaluating its activity against diverse cancer cell lines In vitro studies reported IC50 values typically ranging from approximately 0 1 to several micromolar concentrations varying according to cell type and experimental conditions Therefore pirarubicin serves as a tool compound in pharmacological studies investigating DNA-topoisomerase inhibitors and therapeutic resistance mechanisms
Catalog No. 50-000-61820