Learn More
Medchemexpress LLC Pirarubicin hydrochloride | 95343-20-7 | MFCD01713157 | 99.1% | 664.10 g/mol | C32H38ClNO12 | 5 MG

Supplier: Medchemexpress LLC HY13725A5MG
Pirarubicin hydrochloride is an anthracycline antineoplastic agent and a topoisomerase II inhibitor supplied as the hydrochloride salt. It is used in oncology research, particularly studies of solid tumors, and is provided as a high-purity solid or as a DMSO solution for laboratory applications.
- High purity 99.14%.
- Topoisomerase II inhibitor mechanism of action.
- Hydrochloride salt in solid form.
- Molecular weight 664.10 g/mol; formula C32H38ClNO12.
- Available in small research pack sizes and in DMSO solution.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.