Learn More
Selleck Chemical LLC Pirarubicin 50mg 72496-41-4
SDP
Supplier:  Selleck Chemical LLC S139350MG
Encompass_Preferred
Pirarubicin is an anthracycline antibiotic, and also a DNA/RNA synthesis inhibitor by intercalating into DNA and interacts with topoisomerase II, used as an antineoplastic agent. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-750-2