Learn More
Medchemexpress LLC Pirenzepine dihydrochloride | 29868-97-1 | 99.8% | 424.32 | 1 G
SDP
Catalog No. 5000386088
Encompass_Preferred
Pirenzepine dihydrochloride is a selective M1 muscarinic acetylcholine receptor antagonist that poorly penetrates the blood-brain barrier. It is used in peptic ulcer research due to its ability to reduce gastric acid secretion and muscle spasm. This compound also exhibits anti-proliferative activity against cancer cells.
  • Selective M1 muscarinic acetylcholine receptor antagonist.
  • Reduces gastric acid secretion.
  • Reduces muscle spasm.
  • Useful in peptic ulcer research.
  • Exhibits anti-proliferative activity against cancer cells.
Catalog No. 50-003-86088 Supplier Medchemexpress LLC Supplier No. HY170371G
Pirenzepine dihydrochloride is a selective M1 muscarinic acetylcholine receptor antagonist that poorly penetrates the blood-brain barrier. It is used in peptic ulcer research due to its ability to reduce gastric acid secretion and muscle spasm. This compound also exhibits anti-proliferative activity against cancer cells.
  • Selective M1 muscarinic acetylcholine receptor antagonist.
  • Reduces gastric acid secretion.
  • Reduces muscle spasm.
  • Useful in peptic ulcer research.
  • Exhibits anti-proliferative activity against cancer cells.