Learn More
Medchemexpress LLC Pirenzepine dihydrochloride | 29868-97-1 | 99.8% | 424.32 | 1 ML
SDP
Catalog No. 5000384671
Encompass_Preferred
Pirenzepine dihydrochloride is a selective M1 mAChR (muscarinic acetylcholine receptor) antagonist with poor penetration of the blood-brain barrier. It reduces gastric acid secretion and muscle spasm, and can be used in peptic ulcers research. Pirenzepine dihydrochloride also shows anti-proliferative activity to cancer cells.
  • Selective M1 mAChR antagonist
  • Poor penetration of the blood-brain barrier
  • Reduces gastric acid secretion
  • Reduces muscle spasm
  • Can be used in peptic ulcers research
  • Shows anti-proliferative activity to cancer cells
Catalog No. 50-003-84671 Supplier Medchemexpress LLC Supplier No. HY1703710MM/1ML
Pirenzepine dihydrochloride is a selective M1 mAChR (muscarinic acetylcholine receptor) antagonist with poor penetration of the blood-brain barrier. It reduces gastric acid secretion and muscle spasm, and can be used in peptic ulcers research. Pirenzepine dihydrochloride also shows anti-proliferative activity to cancer cells.
  • Selective M1 mAChR antagonist
  • Poor penetration of the blood-brain barrier
  • Reduces gastric acid secretion
  • Reduces muscle spasm
  • Can be used in peptic ulcers research
  • Shows anti-proliferative activity to cancer cells