Learn More
Selleck Chemical LLC Pirenzepine dihydrochloride S3662-25mg
SDP
Catalog No. 503108913
Encompass_Preferred
Pirenzepine is an antimuscarinic agent which inhibits gastric acid secretion at lower doses than are required to affect gastrointestinal motility salivary central nervous system cardiovascular ocular and urinary functions
Catalog No. 50-310-8913 Supplier Selleck Chemical LLC Supplier No. S366225MG
Pirenzepine is an antimuscarinic agent which inhibits gastric acid secretion at lower doses than are required to affect gastrointestinal motility salivary central nervous system cardiovascular ocular and urinary functions