Learn More
Abcam Pirenzepine dihydrochloride, M1 antagonist, 500MG
Catalog No. 502707836
Encompass_Preferred
MW 424.3 Da, Purity >98%. Selective M1 receptor antagonist. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7836 Supplier Abcam Supplier No. AB120153500MG
MW 424.3 Da, Purity >98%. Selective M1 receptor antagonist. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement