Learn More
Medchemexpress LLC Pirepemat fumarate | 2251806-70-7 | 98.3% | 329.30 | 50 MG
SDP
Catalog No. 5000351554
Encompass_Preferred
Pirepemat fumarate is a cortical-preferring catecholamine- and cognition-promoting agent. It is utilized in the study of Parkinson's disease and is intended for research use only.
  • Highest in vitro affinities for 5-HT and NA-related targets.
  • Reverses deficits resulting from hypomonoaminergic function in vivo.
  • Induces dose-dependent and regio-selective alterations in brain monoamine transmission indices and gene expression.
Catalog No. 50-003-51554 Supplier Medchemexpress LLC Supplier No. HY137447A50MG
Pirepemat fumarate is a cortical-preferring catecholamine- and cognition-promoting agent. It is utilized in the study of Parkinson's disease and is intended for research use only.
  • Highest in vitro affinities for 5-HT and NA-related targets.
  • Reverses deficits resulting from hypomonoaminergic function in vivo.
  • Induces dose-dependent and regio-selective alterations in brain monoamine transmission indices and gene expression.