Learn More
Medchemexpress LLC Pirepemat fumarate | 2251806-70-7 | 98.3% | 329.30 | C15H17F2NO5 | 5 MG
SDP
Catalog No. 5000223394
Encompass_Preferred
Pirepemat fumarate is a cortical-preferring catecholamine- and cognition-promoting research compound used in preclinical and translational studies of Parkinson's disease. Supplied as a solid powder with high reported purity, it is intended for laboratory research applications and is available in small pack sizes or as a standardized solution for screening.
Catalog No. 50-002-23394 Supplier Medchemexpress LLC Supplier No. HY137447A5MG
Pirepemat fumarate is a cortical-preferring catecholamine- and cognition-promoting research compound used in preclinical and translational studies of Parkinson's disease. Supplied as a solid powder with high reported purity, it is intended for laboratory research applications and is available in small pack sizes or as a standardized solution for screening.