Learn More
Medchemexpress LLC Pirepemat fumarate | 2251806-70-7 | 98.3% | C15H17F2NO5 | 10MG
SDP
Catalog No. 5000260129
Encompass_Preferred
Pirepemat fumarate is the fumarate salt of pirepemat (IRL752), a cortical-preferring catecholamine- and cognition-promoting research compound investigated for reducing falls and improving cognitive and motor function in Parkinson's disease. The compound is provided as a stable fumarate salt and is used in preclinical and clinical research to modulate cortical neurotransmitter signaling.
  • Fumarate salt for improved chemical stability.
  • Cortical-preferring catecholamine and cognition-promoting activity.
  • Investigated for reduction of falls and improvement of balance in Parkinson's disease.
  • Shown to enhance cortical dopamine, acetylcholine, and norepinephrine transmission.
  • Supplied in solid and DMSO solution formats for flexible dosing and formulation.
  • High purity suitable for research applications.
Catalog No. 50-002-60129 Supplier Medchemexpress LLC Supplier No. HY137447A10MG
Pirepemat fumarate is the fumarate salt of pirepemat (IRL752), a cortical-preferring catecholamine- and cognition-promoting research compound investigated for reducing falls and improving cognitive and motor function in Parkinson's disease. The compound is provided as a stable fumarate salt and is used in preclinical and clinical research to modulate cortical neurotransmitter signaling.
  • Fumarate salt for improved chemical stability.
  • Cortical-preferring catecholamine and cognition-promoting activity.
  • Investigated for reduction of falls and improvement of balance in Parkinson's disease.
  • Shown to enhance cortical dopamine, acetylcholine, and norepinephrine transmission.
  • Supplied in solid and DMSO solution formats for flexible dosing and formulation.
  • High purity suitable for research applications.