Learn More
Medchemexpress LLC PIRFENIDONE 10 MM 1 ML IN DMSO
SDP
Supplier:  Medchemexpress LLC HYB067310MM 1ML IN DMSO
Encompass_Preferred
Pirfenidone (AMR69) is an antifibrotic agent that attenuates CCL2 and CCL12 production in fibrocyte cells. Pirfenidone has growth-inhibitory effect and reduces TGF-β2 protein levels in human glioma cell lines. Pirfenidone also has anti-inflammatory activities.
Catalog No. NC2480848