Learn More
Selleck Chemical LLC Pirfenidone 10mg 53179-13-8 S-7701,AMR-69
SDP
Catalog No. 501363184
Encompass_Preferred
Pirfenidone (S-7701, AMR-69) is an inhibitor for TGF-? production and TGF-? stimulated collagen production, reduces production of TNF-? and IL-1?, and also has anti-fibrotic and anti-inflammatory properties. Pirfenidone attenuates chemokine (CC motif) ligand-2 (CCL2) and CCL12 production with anti-fibrotic activity. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3184 Supplier Selleck Chemical LLC Supplier No. S290710MG
Pirfenidone (S-7701, AMR-69) is an inhibitor for TGF-? production and TGF-? stimulated collagen production, reduces production of TNF-? and IL-1?, and also has anti-fibrotic and anti-inflammatory properties. Pirfenidone attenuates chemokine (CC motif) ligand-2 (CCL2) and CCL12 production with anti-fibrotic activity. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.