Learn More
Medchemexpress LLC HY-B0673 1g Medchemexpress, Pirfenidone CAS:53179-13-8 Purity:>98%
SDP
Catalog No. 502029402
Encompass_Preferred
Medchemexpress, HY-B0673 1g Pirfenidone CAS:53179-13-8 Pirfenidone (AMR69) is an antifibrotic agent that attenuates CCL2 and CCL12 production in fibrocyte cells. Pirfenidone has growth-inhibitory effect and reduces TGF-β2 protein levels in human glioma cell lines. Pirfenidone also has anti-inflammatory activities[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9402 Supplier Medchemexpress LLC Supplier No. HYB06731G
Medchemexpress, HY-B0673 1g Pirfenidone CAS:53179-13-8 Pirfenidone (AMR69) is an antifibrotic agent that attenuates CCL2 and CCL12 production in fibrocyte cells. Pirfenidone has growth-inhibitory effect and reduces TGF-β2 protein levels in human glioma cell lines. Pirfenidone also has anti-inflammatory activities[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.