Learn More
Medchemexpress LLC HY-B0673 5g , Pirfenidone AMR69 CAS:53179-13-8 Purity:98%
SDP
Catalog No. 501492185
Encompass_Preferred
Medchemexpress, HY-B0673 5g Pirfenidone AMR69 CAS:53179-13-8 Formula:C12H11NO TGF-β 2 Purity:98% Pirfenidone leads to a reduction of TGF-β 2 mRNA levels and of the mature TGF-β 2 protein due to decreased expression and direct inhibition of the TGF-β pro-protein convertase furin. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2185 Supplier Medchemexpress LLC Supplier No. HYB06735G
Medchemexpress, HY-B0673 5g Pirfenidone AMR69 CAS:53179-13-8 Formula:C12H11NO TGF-β 2 Purity:98% Pirfenidone leads to a reduction of TGF-β 2 mRNA levels and of the mature TGF-β 2 protein due to decreased expression and direct inhibition of the TGF-β pro-protein convertase furin. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.