Learn More
Medchemexpress LLC Pirfenidone-d5 | 1020719-62-3 | 98.88% | 190.25 | 1 MG
SDP
Catalog No. 5000365553
Encompass_Preferred
Pirfenidone-d5 (AMR69-d5) is a deuterium labeled Pirfenidone, an antifibrotic agent. It attenuates CCL2 and CCL12 production in fibrocyte cells and reduces TGF-β2 protein levels in human glioma cell lines. It also exhibits anti-inflammatory activities and can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Acts as an antifibrotic agent
  • Attenuates CCL2 and CCL12 production in fibrocyte cells
  • Reduces TGF-β2 protein levels in human glioma cell lines
  • Exhibits anti-inflammatory activities
  • Can be used as a tracer for quantitative analysis
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes may affect pharmacokinetic and metabolic profiles
Catalog No. 50-003-65553 Supplier Medchemexpress LLC Supplier No. 1MGHYB0673S
Add to Cart
Add to Cart
Pirfenidone-d5 (AMR69-d5) is a deuterium labeled Pirfenidone, an antifibrotic agent. It attenuates CCL2 and CCL12 production in fibrocyte cells and reduces TGF-β2 protein levels in human glioma cell lines. It also exhibits anti-inflammatory activities and can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Acts as an antifibrotic agent
  • Attenuates CCL2 and CCL12 production in fibrocyte cells
  • Reduces TGF-β2 protein levels in human glioma cell lines
  • Exhibits anti-inflammatory activities
  • Can be used as a tracer for quantitative analysis
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes may affect pharmacokinetic and metabolic profiles