Learn More
Medchemexpress LLC Pirfenidone-d5 | 1020719-62-3 | 98.9% | 5 MG
SDP
Catalog No. 5000407711
Encompass_Preferred
Pirfenidone-d5 (AMR69-d5) is a deuterium labeled Pirfenidone. It acts as an antifibrotic agent by attenuating CCL2 and CCL12 production in fibrocyte cells. This compound also exhibits a growth-inhibitory effect, reducing TGF-β2 protein levels in human glioma cell lines, and possesses anti-inflammatory activities. It is intended for research use only and can serve as a tracer or an internal standard for quantitative analysis.
  • Used as a tracer for quantitative analysis
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Attenuates CCL2 and CCL12 production in fibrocyte cells
  • Exhibits a growth-inhibitory effect
  • Reduces TGF-β2 protein levels in human glioma cell lines
  • Possesses anti-inflammatory activities
Catalog No. 50-004-07711 Supplier Medchemexpress LLC Supplier No. HYB0673S5MG
Pirfenidone-d5 (AMR69-d5) is a deuterium labeled Pirfenidone. It acts as an antifibrotic agent by attenuating CCL2 and CCL12 production in fibrocyte cells. This compound also exhibits a growth-inhibitory effect, reducing TGF-β2 protein levels in human glioma cell lines, and possesses anti-inflammatory activities. It is intended for research use only and can serve as a tracer or an internal standard for quantitative analysis.
  • Used as a tracer for quantitative analysis
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Attenuates CCL2 and CCL12 production in fibrocyte cells
  • Exhibits a growth-inhibitory effect
  • Reduces TGF-β2 protein levels in human glioma cell lines
  • Possesses anti-inflammatory activities