Learn More
Medchemexpress LLC Pirfenidone-d5 | 1020719-62-3 | MFCD07369537 | 98.9% | 190.25 g/mol | C12H6D5NO | 10 MG
SDP
Catalog No. 5000298079
Encompass_Preferred
Pirfenidone-d5 is the deuterium-labeled isotopologue of pirfenidone used as an analytical reference and internal standard in quantitative mass spectrometry and pharmacokinetic studies. It provides a chemically similar labeled compound for accurate quantitation of pirfenidone in biological and biochemical assays.
  • Deuterium-labeled internal standard for LC-MS/MS assays
  • High purity (~98.9%) suitable for analytical use
  • Molecular weight 190.25 g/mol; formula C12H6D5NO
  • Available as a 10 mg vial for laboratory use
  • Suitable for pharmacokinetic and metabolic studies
Catalog No. 50-002-98079 Supplier Medchemexpress LLC Supplier No. HYB0673S10MG
Pirfenidone-d5 is the deuterium-labeled isotopologue of pirfenidone used as an analytical reference and internal standard in quantitative mass spectrometry and pharmacokinetic studies. It provides a chemically similar labeled compound for accurate quantitation of pirfenidone in biological and biochemical assays.
  • Deuterium-labeled internal standard for LC-MS/MS assays
  • High purity (~98.9%) suitable for analytical use
  • Molecular weight 190.25 g/mol; formula C12H6D5NO
  • Available as a 10 mg vial for laboratory use
  • Suitable for pharmacokinetic and metabolic studies