Learn More
Abcam Pirfenidone, orally active anti-inflammatory antifibrotic agent, 50MG
Catalog No. 502706627
Encompass_Preferred
MW 185.22 Da, Purity >99%. Orally active anti-inflammatory, antifibrotic and antiproliferative agent. Blood-brain barrier permeable. Shown to modulate cytokines TGF-β and TNF-α.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6627 Supplier Abcam Supplier No. AB14142550MG
Add to Cart
Add to Cart
MW 185.22 Da, Purity >99%. Orally active anti-inflammatory, antifibrotic and antiproliferative agent. Blood-brain barrier permeable. Shown to modulate cytokines TGF-β and TNF-α.
The product is subject to the following: Abcam Restricted Use Statement