Learn More
Medchemexpress LLC Piribedil-d8 | 1398044-45-5 | 99.8% | 1 MG
SDP
Catalog No. 5000342437
Encompass_Preferred
Piribedil-d8 is a deuterium-labeled analog of Piribedil, an antiparkinsonian agent known for its activity as a dopamine D2 and D3 agonist and α2-adrenoceptors antagonist. This compound serves as a valuable tracer and internal standard for quantitative analysis in various analytical techniques, including NMR, GC-MS, and LC-MS.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Acts as a dopamine D2 and D3 agonist
  • Serves as an α2-adrenoceptors antagonist
  • Inhibits MLL1 methyltransferase activity
  • Potential for research in Parkinson's disease, circulatory disorders, and cancers
Catalog No. 50-003-42437 Supplier Medchemexpress LLC Supplier No. HY12707S1MG
Piribedil-d8 is a deuterium-labeled analog of Piribedil, an antiparkinsonian agent known for its activity as a dopamine D2 and D3 agonist and α2-adrenoceptors antagonist. This compound serves as a valuable tracer and internal standard for quantitative analysis in various analytical techniques, including NMR, GC-MS, and LC-MS.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Acts as a dopamine D2 and D3 agonist
  • Serves as an α2-adrenoceptors antagonist
  • Inhibits MLL1 methyltransferase activity
  • Potential for research in Parkinson's disease, circulatory disorders, and cancers