Learn More
Medchemexpress LLC Piribedil-d8 | 1398044-45-5 | MFCD28900715 | 99.8% | 306.39 | C16H10D8N4O2 | 5 MG
SDP
Catalog No. 5000264091
Encompass_Preferred
Piribedil-d8 is the deuterium-labeled analogue of piribedil used primarily as an internal standard or tracer for quantitative analytical methods. Supplied as a white to off-white solid with high isotopic and chemical purity, it is intended for use in NMR, GC-MS, and LC-MS assays and pharmacokinetic studies. Store and handle according to manufacturer recommendations.
  • Deuterium-labeled internal standard for accurate quantitation
  • High purity suitable for analytical and bioanalytical workflows
  • Soluble in DMSO for convenient stock solution preparation
  • Stable when stored under recommended temperature conditions
  • Applicable as tracer and internal standard in mass spectrometry and NMR analyses
Catalog No. 50-002-64091 Supplier Medchemexpress LLC Supplier No. HY12707S5MG
Piribedil-d8 is the deuterium-labeled analogue of piribedil used primarily as an internal standard or tracer for quantitative analytical methods. Supplied as a white to off-white solid with high isotopic and chemical purity, it is intended for use in NMR, GC-MS, and LC-MS assays and pharmacokinetic studies. Store and handle according to manufacturer recommendations.
  • Deuterium-labeled internal standard for accurate quantitation
  • High purity suitable for analytical and bioanalytical workflows
  • Soluble in DMSO for convenient stock solution preparation
  • Stable when stored under recommended temperature conditions
  • Applicable as tracer and internal standard in mass spectrometry and NMR analyses