Learn More
Abcam Piribedil dihydrochloride, D2/D3 dopamine receptor agonist, 50MG
Catalog No. 502704616
Encompass_Preferred
MW 371.27 g/mol, Purity >99%. D₂/D₃ dopamine receptor agonist. D₃ selectivity demonstrated in vitro. Improves attentional dysfunction in combination with L-DOPA.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4616 Supplier Abcam Supplier No. AB14643250MG
MW 371.27 g/mol, Purity >99%. D₂/D₃ dopamine receptor agonist. D₃ selectivity demonstrated in vitro. Improves attentional dysfunction in combination with L-DOPA.
The product is subject to the following: Abcam Restricted Use Statement