Learn More
Medchemexpress LLC Pirimiphos-methyl | 29232-93-7 | 98.39% | 250 MG
SDP
Catalog No. 5000414337
Encompass_Preferred
Pirimiphos-methyl is an organophosphorus insecticide and acaricide that inhibits AChE in target organisms. It is commonly used for the prevention and control of beetles, snout beetles, moths, and Ephestia cautella during the storage of agricultural grains. It is intended for research use only.
  • Organophosphorus insecticide and acaricide.
  • Inhibits AChE in target organisms.
  • Used for prevention and control of various pests in stored agricultural grains.
  • Available in various sizes, including 10 mM * 1 mL in DMSO (liquid + solvent and solution forms), 100 mg, 250 mg, 500 mg liquid, and larger quantities by quote.
  • Soluble in DMSO.
Catalog No. 50-004-14337 Supplier Medchemexpress LLC Supplier No. 250MGHYB1881
Pirimiphos-methyl is an organophosphorus insecticide and acaricide that inhibits AChE in target organisms. It is commonly used for the prevention and control of beetles, snout beetles, moths, and Ephestia cautella during the storage of agricultural grains. It is intended for research use only.
  • Organophosphorus insecticide and acaricide.
  • Inhibits AChE in target organisms.
  • Used for prevention and control of various pests in stored agricultural grains.
  • Available in various sizes, including 10 mM * 1 mL in DMSO (liquid + solvent and solution forms), 100 mg, 250 mg, 500 mg liquid, and larger quantities by quote.
  • Soluble in DMSO.