Learn More
Cayman Chemical PIrInIxIc ACD AmInothIazl 10mg
SDP
Catalog No. 502971402
Encompass_Preferred
A dual inhibitor of mPGES-1 and 5-LO (IC50s = 0.4 and 0.2 µM, respectively); reduces the synthesis of PGE2 and LTC4 during zymosan-induced peritonitis in mice, resulting in a significantly diminished inflammatory response
Catalog No. 50-297-1402 Supplier Cayman Chemical Supplier No. 1548310 MG
A dual inhibitor of mPGES-1 and 5-LO (IC50s = 0.4 and 0.2 µM, respectively); reduces the synthesis of PGE2 and LTC4 during zymosan-induced peritonitis in mice, resulting in a significantly diminished inflammatory response