Learn More
Medchemexpress LLC HY-16995 10mg Medchemexpress, Pirinixic acid CAS:50892-23-4 Purity:>98%
SDP
Catalog No. 502081922
Encompass_Preferred
Medchemexpress, HY-16995 10mg Pirinixic acid CAS:50892-23-4 Pirinixic acid (Wy-14643) is a potent agonist of PPARα, with EC50s of 0.63 μM, 32 μM for murine PPARα and PPARγ, and 5.0 μM, 60 μM, 35 μM for human PPARα, PPARγ and PPARδ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1922 Supplier Medchemexpress LLC Supplier No. HY1699510MG
Medchemexpress, HY-16995 10mg Pirinixic acid CAS:50892-23-4 Pirinixic acid (Wy-14643) is a potent agonist of PPARα, with EC50s of 0.63 μM, 32 μM for murine PPARα and PPARγ, and 5.0 μM, 60 μM, 35 μM for human PPARα, PPARγ and PPARδ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.