Learn More
Medchemexpress LLC Pirinixic acid | 50892-23-4 | 323.80 | 250 MG
SDP
Catalog No. 5000395410
Encompass_Preferred
Pirinixic acid, also known as Wy-14643, is a potent agonist of peroxisome proliferator-activated receptor alpha (PPARα). It is widely used in research concerning metabolic diseases and inflammation. This compound has demonstrated significant activity against various PPAR subtypes in both murine and human models, affecting cellular processes related to inflammation and energy metabolism.
  • Functions as a potent agonist of PPARα.
  • Suitable for research in metabolic disease and inflammation/immunology.
  • Decreases hepatic injury and lipid peroxidation.
  • Enhances PPAR-α protein expression in synovial fibroblasts.
  • Inhibits NO and PGE2 production in LPS-stimulated synovial fibroblasts.
  • Available as a solid for long-term storage.
Catalog No. 50-003-95410 Supplier Medchemexpress LLC Supplier No. 250MGHY16995
Pirinixic acid, also known as Wy-14643, is a potent agonist of peroxisome proliferator-activated receptor alpha (PPARα). It is widely used in research concerning metabolic diseases and inflammation. This compound has demonstrated significant activity against various PPAR subtypes in both murine and human models, affecting cellular processes related to inflammation and energy metabolism.
  • Functions as a potent agonist of PPARα.
  • Suitable for research in metabolic disease and inflammation/immunology.
  • Decreases hepatic injury and lipid peroxidation.
  • Enhances PPAR-α protein expression in synovial fibroblasts.
  • Inhibits NO and PGE2 production in LPS-stimulated synovial fibroblasts.
  • Available as a solid for long-term storage.