Learn More
Cayman Chemical Piriprost (potassium salt), Purity: ≥98%, CAS: 88851-62-1, 50 mg
SDP
Catalog No. 502965178
Encompass_Preferred
Piriprost is a structural analog of PGI2 with low IP receptor-mediated activity. Piriprost inhibits 5-LO with an IC50 value around 100 µM, as measured by the release of 5-HETE from cultured myometrial cells.{994} Piriprost inhibits the release of histamine and leukotrienes from isolated porcine lung cells with an IC50 value of 0.11 µM.{4708}
Catalog No. 50-296-5178 Supplier Cayman Chemical Supplier No. 1822850 MG
Piriprost is a structural analog of PGI2 with low IP receptor-mediated activity. Piriprost inhibits 5-LO with an IC50 value around 100 µM, as measured by the release of 5-HETE from cultured myometrial cells.{994} Piriprost inhibits the release of histamine and leukotrienes from isolated porcine lung cells with an IC50 value of 0.11 µM.{4708}