Learn More
Medchemexpress LLC Pirlimycin hydrochloride | 78822-40-9 | MFCD27952783 | 99.1% | 447.42 g·mol⁻¹ | C17H32Cl2N2O5S | 1 MG
SDP
Catalog No. 5000240522
Encompass_Preferred
Pirlimycin hydrochloride is a lincosamide-class antibiotic supplied as the hydrochloride salt for research use. It inhibits bacterial protein synthesis by binding the 50S ribosomal subunit and is primarily active against Gram-positive organisms. The material is provided as a solid at high purity for analytical and laboratory applications.
  • Lincosamide antibiotic.
  • Inhibits bacterial protein synthesis via 50S ribosomal binding.
  • Primarily active against Gram-positive bacteria.
  • High purity (99.1%).
  • Supplied as a solid suitable for analytical use.
  • Available in small-scale quantities for research.
Catalog No. 50-002-40522 Supplier Medchemexpress LLC Supplier No. HY106597A1MG
Pirlimycin hydrochloride is a lincosamide-class antibiotic supplied as the hydrochloride salt for research use. It inhibits bacterial protein synthesis by binding the 50S ribosomal subunit and is primarily active against Gram-positive organisms. The material is provided as a solid at high purity for analytical and laboratory applications.
  • Lincosamide antibiotic.
  • Inhibits bacterial protein synthesis via 50S ribosomal binding.
  • Primarily active against Gram-positive bacteria.
  • High purity (99.1%).
  • Supplied as a solid suitable for analytical use.
  • Available in small-scale quantities for research.