Learn More
Cayman Chemical Pirlindole-d4 (hydrochloride), Purity: ≥99% deuterated forms (d1-d4), CAS: 1801617-88-8, 1 mg
SDP
Catalog No. 502948459
Encompass_Preferred
An internal standard for the quantification of pirlindole by GC- or LC-MS
Catalog No. 50-294-8459 Supplier Cayman Chemical Supplier No. 333751 MG
An internal standard for the quantification of pirlindole by GC- or LC-MS